Stampa…

Benziodarone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-Ethyl-3-(4-hydroxy-3,5-diiodobenzoyl)benzofuran

E1290-1G E1290-5G
TCIAE1290-1GEA 77 EUR
TCIAE1290-1G TCIAE1290-5G
Benziodarone ≥98.0% (by GC)
Benziodarone
Formula: C₁₇H₁₂I₂O₃
Peso molecolare: 518,09 g/mol
MDL Number: MFCD00868246
CAS Number: 68-90-6
EINECS: 200-695-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,01083

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 164.0 to 168.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR