Stampa…
Esculetin ≥99.0% (by HPLC, titration analysis) matrix for MALDI-TOF/MS
Fornitore: TCI EUROPE
Attenzione
Synonyms: 6,7-Dihydroxycoumarin, 6,7-Dihydroxy-2H-chromen-2-one
E1287-200MG E1287-1G
TCIAE1287-200MGEA 28 EUR
TCIAE1287-200MG TCIAE1287-1G
Esculetin ≥99.0% (by HPLC, titration analysis) matrix for MALDI-TOF/MS
Esculetin
Formula: C₉H₆O₄
Peso molecolare: 178,14 g/mol
Punto di fusione: 273°C
MDL Number: MFCD00006874
CAS Number: 305-01-1
EINECS: 206-161-5
Merck Index: 14,03697
Visualizza i codici
Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %