Stampa…

Ethyl-5-bromo-2-furoate ≥97.0% (by GC)

Fornitore: TCI

Synonyms: Ethyl 5-bromofuran-2-carboxylate, Ethyl 5-Bromo-2-furoate, Ethyl 5-Bromo-2-furancarboxylate, 5-Bromo-2-furoic acid ethyl ester, 5-Bromo-2-furancarboxylic acid ethyl ester

E1285-1G E1285-5G
TCIAE1285-1GEA 79 EUR
TCIAE1285-1G TCIAE1285-5G
Ethyl-5-bromo-2-furoate ≥97.0% (by GC)
Ethyl-5-bromo-2-furoate

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR