Stampa…

Ethyl(dimethyl)(2-phenylethyl)ammonium bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ethyl(dimethyl)phenethylammonium bis(trifluoromethanesulphonyl)imide

E1283-1G E1283-5G
TCIAE1283-1GEA 47 EUR
TCIAE1283-1G TCIAE1283-5G
Ethyl(dimethyl)(2-phenylethyl)ammonium bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)
Ethyl(dimethyl)(2-phenylethyl)ammonium bis(trifluoromethanesulphonyl)imide
Formula: C₁₄H₂₀F₆N₂O₄S₂
Peso molecolare: 458,43 g/mol
CAS Number: 1804970-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41f1fc856dd63055a4c41b4e2d2a52dd940abcff&vid=41f1fc856dd63055a4c41b4e2d2a52dd940abcff&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to lump
Purity (HPLC) min. 96.0 area%
Purity (Ion exchange titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR