Ethyl-4-(1-piperazinyl)benzoate ≥98.0% (by GC)
Fornitore: TCI
Synonyms:
Ethyl 4-(piperazin-1-yl)benzoate
Formula:
C₁₃H₁₈N₂O₂ Peso molecolare: 234.30 g/mol |
MDL Number:
MFCD04973340 CAS Number: 80518-57-6 |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 100.0 to 104.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...