Stampa…

Ethyl (3-Hydroxyphenyl)acetate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ethyl 2-(3-hydroxyphenyl)acetate

E1242-1G E1242-5G
TCIAE1242-1GEA 32 EUR
TCIAE1242-1G TCIAE1242-5G
Ethyl (3-Hydroxyphenyl)acetate ≥98.0% (by GC)
Ethyl (3-Hydroxyphenyl)acetate
Formula: C₁₀H₁₂O₃
Peso molecolare: 180,20 g/mol
Punto di ebollizione: 179°C
MDL Number: MFCD07369387
CAS Number: 22446-38-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce7a1d28f79392b425fa1b52529d500652981ccf&vid=ce7a1d28f79392b425fa1b52529d500652981ccf&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR