Stampa…
Ethyl-2-aminothiazole-4-carboxylate ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Attenzione
Synonyms: Ethyl 2-aminothiazole-4-carboxylate
E1215-5G E1215-25G
TCIAE1215-5GEA 53 EUR
TCIAE1215-5G TCIAE1215-25G
Ethyl-2-aminothiazole-4-carboxylate ≥98.0% (by GC, titration analysis)
Ethyl-2-aminothiazole-4-carboxylate
Formula: C₆H₈N₂O₂S
Peso molecolare: 172,21 g/mol
MDL Number: MFCD00619079
CAS Number: 5398-36-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbb3d195c21c75974fd4d5bbb6dc4a9568baebf0&vid=fbb3d195c21c75974fd4d5bbb6dc4a9568baebf0&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 178.0 to 182.0 deg-C