Stampa…

4-(Ethoxycarbonyl)-3-fluorophenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Ethoxycarbonyl-3-fluorobenzeneboronic acid, 4-Ethoxycarbonyl-3-fluorophenylboronic acid

E1207-1G E1207-5G
TCIAE1207-1GEA 70 EUR
TCIAE1207-1G TCIAE1207-5G
4-(Ethoxycarbonyl)-3-fluorophenylboronic acid (contains varying amounts of Anhydride)
4-(Ethoxycarbonyl)-3-fluorophenylboronic acid
Formula: C₉H₁₀BFO₄
Peso molecolare: 211,98 g/mol
MDL Number: MFCD06656274
CAS Number: 874288-38-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7350b2d1bf473035153bc35e3111af7556fd439b&vid=7350b2d1bf473035153bc35e3111af7556fd439b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR