Stampa…

2-Ethoxy-4-fluorophenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

E1204-1G E1204-5G
TCIAE1204-1GEA 56 EUR
TCIAE1204-1G TCIAE1204-5G
2-Ethoxy-4-fluorophenylboronic acid (contains varying amounts of Anhydride)
2-Ethoxy-4-fluorophenylboronic acid
Formula: C₈H₁₀BFO₃
Peso molecolare: 183,97 g/mol
MDL Number: MFCD03095001
CAS Number: 480438-58-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d999beb96194068c36f6676e9b6d9e765ba21ad7&vid=d999beb96194068c36f6676e9b6d9e765ba21ad7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 96.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR