4-(trans-4-Ethylcyclohexyl)cyclohexanone ≥98.0% (by GC)
Fornitore: TCI EUROPE
E1197-5G
E1197-25G
TCIAE1197-5GEA
70
EUR
TCIAE1197-5G
TCIAE1197-25G
4-(trans-4-Ethylcyclohexyl)cyclohexanone ≥98.0% (by GC)
4-(trans-4-Ethylcyclohexyl)cyclohexanone
Formula:
C₁₄H₂₄O Peso molecolare: 208,35 g/mol |
MDL Number:
MFCD09750931 CAS Number: 150763-46-5 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...