Stampa…

Ethyl-2,6-dichloropyridine-3-carboxylate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ethyl 2,6-Dichloropyridine-3-carboxylate, 2,6-Dichloropyridine-3-carboxylic acid ethyl ester, 2,6-Dichloronicotinic acid ethyl ester, Ethyl 2,6-Dichloronicotinate

E1194-1G E1194-5G
TCIAE1194-1GEA 56 EUR
TCIAE1194-1G TCIAE1194-5G
Ethyl-2,6-dichloropyridine-3-carboxylate ≥98.0% (by GC)
Ethyl-2,6-dichloropyridine-3-carboxylate
Formula: C₈H₇Cl₂NO₂
Peso molecolare: 220,06 g/mol
MDL Number: MFCD09757510
CAS Number: 58584-86-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR