Stampa…

Ethyl potassium oxalate ≥95.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Potassium 2-ethoxy-2-oxoacetate

E1184-5G E1184-25G
TCIAE1184-5GEA 78 EUR
TCIAE1184-5G TCIAE1184-25G
Ethyl potassium oxalate ≥95.0% (by titrimetric analysis)
Ethyl potassium oxalate
Formula: C₄H₅KO₄
Peso molecolare: 156,178 g/mol
MDL Number: MFCD00045861
CAS Number: 1906-57-6
EINECS: 217-606-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR