Stampa…

4-Ethoxy-2-methylbenzeneboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Ethoxy-2-methylphenylboronic acid, 4-Ethoxy-2-methylbenzeneboronic acid, B-(4-Ethoxy-2-methylphenyl)boronic acid

E1173-1G E1173-5G
TCIAE1173-1GEA 42 EUR
TCIAE1173-1G TCIAE1173-5G
4-Ethoxy-2-methylbenzeneboronic acid (contains varying amounts of Anhydride)
4-Ethoxy-2-methylbenzeneboronic acid
Formula: C₉H₁₃BO₃
Peso molecolare: 180,01 g/mol
MDL Number: MFCD04115659
CAS Number: 313545-31-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR