Stampa…

(4-Ethoxyphenyl)urea ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-(4-Ethoxyphenyl)urea

E1171-200MG E1171-1G
TCIAE1171-200MGEA 53 EUR
TCIAE1171-200MG TCIAE1171-1G
(4-Ethoxyphenyl)urea ≥98.0% (by HPLC, total nitrogen)
(4-Ethoxyphenyl)urea
Formula: C₉H₁₂N₂O₂
Peso molecolare: 180,21 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00025431
CAS Number: 150-69-6
EINECS: 205-767-7
Merck Index: 14,03464

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 169.0 to 173.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR