2,3-Dihydrothieno[3,4-b]-1,4-dioxin-5-carbaldehyde ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
2,3-Dihydrothieno[3,4-b][1,4]dioxine-5-carbaldehyde, 3,4-Ethylenedioxythiophene-2-carboxaldehyde, 2,3-Dihydrothieno[3,4-b][1,4]dioxine-5-carboxaldehyde
E1150-1G
E1150-5G
TCIAE1150-1GEA
60
EUR
TCIAE1150-1G
TCIAE1150-5G
2,3-Dihydrothieno[3,4-b]-1,4-dioxin-5-carbaldehyde ≥98.0% (by GC)
2,3-Dihydrothieno[3,4-b]-1,4-dioxin-5-carbaldehyde
Formula:
C₇H₆O₃S Peso molecolare: 170,19 g/mol |
MDL Number:
MFCD01651766 CAS Number: 204905-77-1 |
Risultati del test di specificazione
Purity (GC) | min. 98.0 % |
Melting point | 139.0 to 143.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...