Stampa…

Ethyl-2-aminonicotinate ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ethyl 2-aminonicotinate

E1146-1G E1146-5G
TCIAE1146-1GEA 23 EUR
TCIAE1146-1G TCIAE1146-5G
Ethyl-2-aminonicotinate ≥98.0% (by GC, titration analysis)
Ethyl-2-aminonicotinate
Formula: C₈H₁₀N₂O₂
Peso molecolare: 166,18 g/mol
Punto di ebollizione: 133°C
MDL Number: MFCD01569452
CAS Number: 13362-26-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR