Stampa…

Etamsylate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 2,5-Dihydroxybenzenesulphonic acid-N-ethylethanamine, Diethylamine 2,5-Dihydroxybenzenesulphonate

E1145-1G E1145-5G
TCIAE1145-1GEA 93 EUR
TCIAE1145-1G TCIAE1145-5G
Etamsylate ≥98.0% (by HPLC, titration analysis)
Etamsylate
Formula: C₁₀H₁₇NO₅S
Peso molecolare: 263,31 g/mol
MDL Number: MFCD00867499
CAS Number: 2624-44-4
EINECS: 220-090-7
Merck Index: 14,03722

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 128.0 to 132.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR