Stampa…

Ethyl-2-amino-5-phenylthiophene-3-carboxylate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ethyl 2-amino-5-phenylthiophene-3-carboxylate

E1139-200MG E1139-1G
TCIAE1139-200MGEA 24 EUR
TCIAE1139-200MG TCIAE1139-1G
Ethyl-2-amino-5-phenylthiophene-3-carboxylate ≥98.0% (by GC)
Ethyl-2-amino-5-phenylthiophene-3-carboxylate
Formula: C₁₃H₁₃NO₂S
Peso molecolare: 247,32 g/mol
MDL Number: MFCD01829801
CAS Number: 4815-34-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 122.0 to 126.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR