Stampa…

Ethyl-3-amino-4-pyridinecarboxylate ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ethyl 3-amino-4-pyridinecarboxylate, Ethyl 3-Aminoisonicotinate, 3-Aminoisonicotinic acid ethyl ester, Ethyl 3-Aminopyridine-4-carboxylate, 3-Aminopyridine-4-carboxylic acid ethyl ester

E1137-1G E1137-5G
TCIAE1137-1GEA 51 EUR
TCIAE1137-1G TCIAE1137-5G
Ethyl-3-amino-4-pyridinecarboxylate ≥98.0% (by GC, titration analysis)
Ethyl-3-amino-4-pyridinecarboxylate
Formula: C₈H₁₀N₂O₂
Peso molecolare: 166,1800 g/mol
MDL Number: MFCD03788270
CAS Number: 14208-83-4

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 65.0 to 69.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR