Stampa…

Ethyl (3-Methoxybenzoyl)acetate ≥98.0% (by GC, titration analysis)

Fornitore: TCI

E1121-1G E1121-5G
TCIAE1121-1GEA 41 EUR
TCIAE1121-1G TCIAE1121-5G
Ethyl (3-Methoxybenzoyl)acetate ≥98.0% (by GC, titration analysis)
Ethyl (3-Methoxybenzoyl)acetate
Formula: C₁₂H₁₄O₄
Punto di ebollizione: 123°C
MDL Number: MFCD00086704
CAS Number: 27834-99-7

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 % (total of isomer)
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR