Stampa…

2-Ethoxy-5-pyridineboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Ethoxy-5-pyridylboronic acid, 6-Ethoxy-3-pyridylboronic acid, 2-Ethoxypyridine-5-boronic acid, (6-Ethoxypyridin-3-yl)boronic acid, 6-Ethoxypyridine-3-boronic acid

E1115-1G E1115-5G
TCIAE1115-1GEA 38 EUR
TCIAE1115-1G TCIAE1115-5G
2-Ethoxy-5-pyridineboronic acid (contains varying amounts of Anhydride)
2-Ethoxy-5-pyridineboronic acid
Formula: C₇H₁₀BNO₃
Peso molecolare: 166,97 g/mol
Punto di fusione: 134°C
MDL Number: MFCD06201032
CAS Number: 612845-44-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR