3-Ethoxy-4-methoxybenzonitrile ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
E1109-5G
E1109-25G
TCIAE1109-5GEA
39
EUR
TCIAE1109-5G
TCIAE1109-25G
3-Ethoxy-4-methoxybenzonitrile ≥98.0% (by GC)
3-Ethoxy-4-methoxybenzonitrile
Formula:
C₁₀H₁₁NO₂ Peso molecolare: 177,20 g/mol |
MDL Number:
MFCD02256160 CAS Number: 60758-86-3 UN: 3439 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 70.0 to 74.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...