Ethyl (2-fluorobenzoyl)acetate ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₁H₁₁FO₃ Punto di ebollizione: 108°C |
MDL Number:
MFCD00000321 CAS Number: 1479-24-9 |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Specific gravity (20/20) | 1.1740 to 1.1860 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...