Stampa…

Ethyl-3-methylindole-2-carboxylate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ethyl 3-methyl-1H-indole-2-carboxylate, 3-Methylindole-2-carboxylic Acid Ethyl Ester

E1090-1G E1090-5G
TCIAE1090-1GEA 76 EUR
TCIAE1090-1G TCIAE1090-5G
Ethyl-3-methylindole-2-carboxylate ≥98.0% (by GC)
Ethyl-3-methylindole-2-carboxylate
Formula: C₁₂H₁₃NO₂
Peso molecolare: 203,24 g/mol
Punto di ebollizione: 195°C
MDL Number: MFCD00225365
CAS Number: 26304-51-8

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 134.0 to 138.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR