Ethyl-1,3-benzothiazole-2-carboxylate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Benzothiazole-2-carboxylic acid ethyl ester, Ethyl 1,3-benzothiazole-2-carboxylate, Ethyl Benzothiazole-2-carboxylate
E1089-5G
E1089-25G
TCIAE1089-5GEA
30
EUR
TCIAE1089-5G
TCIAE1089-25G
Ethyl-1,3-benzothiazole-2-carboxylate ≥98.0% (by GC)
Ethyl-1,3-benzothiazole-2-carboxylate
Formula:
C₁₀H₉NO₂S Peso molecolare: 207,25 g/mol |
MDL Number:
MFCD00848360 CAS Number: 32137-76-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 68.0 to 72.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...