Stampa…

N,N'-Ethylenebisacrylamide ≥97.0% (by GC)

Fornitore: TCI
Attenzione

E1086-5G E1086-25G
TCIAE1086-5GEA 148 EUR
TCIAE1086-5G TCIAE1086-25G
N,N'-Ethylenebisacrylamide ≥97.0% (by GC)
N,N'-Ethylenebisacrylamide
Formula: C₈H₁₂N₂O₂
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00042935
CAS Number: 2956-58-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Bromination Method) min. 95.0 %
Melting point 143.0 to 147.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR