Stampa…

Ethyl thiophene-3-carboxylate ≥95.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Ethyl 3-Thiophenecarboxylate, 3-Thiophenecarboxylic acid ethyl ester

E1082-1G E1082-5G
TCIAE1082-1GEA 25 EUR
TCIAE1082-1G TCIAE1082-5G
Ethyl thiophene-3-carboxylate ≥95.0% (by GC)
Ethyl thiophene-3-carboxylate
Formula: C₇H₈O₂S
Peso molecolare: 156,20 g/mol
Punto di ebollizione: 80°C
MDL Number: MFCD02179226
CAS Number: 5751-80-4
EINECS: 227-275-1

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR