Stampa…

Ethyl 3-(4-Cyanophenyl)-3-oxopropionate ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ethyl 3-(4-cyanophenyl)-3-oxopropanoate, 3-(4-Cyanophenyl)-3-oxopropionic acid Ethyl ester

E1080-200MG E1080-1G
TCIAE1080-200MGEA 81 EUR
TCIAE1080-200MG TCIAE1080-1G
Ethyl 3-(4-Cyanophenyl)-3-oxopropionate ≥98.0% (by GC, titration analysis)
Ethyl 3-(4-Cyanophenyl)-3-oxopropionate
Formula: C₁₂H₁₁NO₃
Peso molecolare: 217,22 g/mol
Punto di ebollizione: 140°C
MDL Number: MFCD00029621
CAS Number: 49744-93-6

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 65.0 to 69.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR