Stampa…

Ethyl-2,4-dimethylpyrrole-3-carboxylate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Ethyl 2,4-dimethyl-1H-pyrrole-3-carboxylate

E1075-1G E1075-5G
TCIAE1075-1GEA 38 EUR
TCIAE1075-1G TCIAE1075-5G
Ethyl-2,4-dimethylpyrrole-3-carboxylate ≥98.0% (by GC)
Ethyl-2,4-dimethylpyrrole-3-carboxylate
Formula: C₉H₁₃NO₂
Peso molecolare: 167,2050 g/mol
Punto di ebollizione: 162°C
MDL Number: MFCD01365811
CAS Number: 2199-51-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 74.0 to 78.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR