Ethyl-2-benzoylpropionate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Ethyl 2-methyl-3-oxo-3-phenylpropanoate, 2-Benzoylpropionic Acid Ethyl Ester
E1071-1G
E1071-5G
TCIAE1071-1GEA
74
EUR
TCIAE1071-1G
TCIAE1071-5G
Ethyl-2-benzoylpropionate ≥98.0% (by GC)
Ethyl-2-benzoylpropionate
Formula:
C₁₂H₁₄O₃ Peso molecolare: 206,24 g/mol Punto di ebollizione: 96°C |
MDL Number:
MFCD00026882 CAS Number: 10488-87-6 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...