Ethyl 3-Oxo-3-(4-chlorophenyl)propionate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Ethyl 3-(4-chlorophenyl)-3-oxo-propionate, 3-Oxo-3-(4-chlorophenyl)propionic acid Ethyl ester
E1059-1G
E1059-5G
TCIAE1059-1GEA
65
EUR
TCIAE1059-1G
TCIAE1059-5G
Ethyl 3-Oxo-3-(4-chlorophenyl)propionate ≥98.0% (by GC)
Ethyl 3-Oxo-3-(4-chlorophenyl)propionate
Formula:
C₁₁H₁₁ClO₃ Peso molecolare: 226,66 g/mol Punto di ebollizione: 123°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00018713 CAS Number: 2881-63-2 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...