Stampa…

5-Ethynyl-2'-deoxyuridine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 5-EdU

E1057-50MG E1057-200MG
TCIAE1057-50MGEA 48 EUR
TCIAE1057-50MG TCIAE1057-200MG
5-Ethynyl-2'-deoxyuridine ≥98.0% (by HPLC, titration analysis)
5-Ethynyl-2'-deoxyuridine
Formula: C₁₁H₁₂N₂O₅
Peso molecolare: 252,23 g/mol
Punto di fusione: 199°C
MDL Number: MFCD01675687
CAS Number: 61135-33-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +21.0 to +25.0 deg (C=0.7, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR