Stampa…

Ethyl 2-(4-Hydroxyphenyl)-4-methylthiazole-5-carboxylate hydrochloride ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: 2-(4-Hydroxyphenyl)-4-methylthiazole-5-carboxylic acid Ethyl ester hydrochloride

E1049-5G E1049-25G
TCIAE1049-5GEA 73 EUR
TCIAE1049-5G TCIAE1049-25G
Ethyl 2-(4-Hydroxyphenyl)-4-methylthiazole-5-carboxylate hydrochloride ≥98.0% (by HPLC, total nitrogen)
Ethyl 2-(4-Hydroxyphenyl)-4-methylthiazole-5-carboxylate hydrochloride
Formula: C₁₃H₁₃NO₃S·HCl
Peso molecolare: 299,77 g/mol
CAS Number: 399017-10-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR