Stampa…

Imidazole-4-carboxylic acid ethyl ester ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ethyl imidazole-4-carboxylate

E1042-1G E1042-5G
TCIAE1042-1GEA 18 EUR
TCIAE1042-1G TCIAE1042-5G
Imidazole-4-carboxylic acid ethyl ester ≥98.0% (by GC, titration analysis)
Imidazole-4-carboxylic acid ethyl ester
Formula: C₆H₈N₂O₂
Peso molecolare: 140,14 g/mol
MDL Number: MFCD07363812
CAS Number: 23785-21-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 158.0 to 163.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR