Ethyl pyrrole-2-carboxylate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Ethyl 1H-pyrrole-2-carboxylate
E1030-5G
E1030-25G
TCIAE1030-5GEA
56
EUR
TCIAE1030-5G
TCIAE1030-25G
Ethyl pyrrole-2-carboxylate ≥98.0% (by GC)
Ethyl pyrrole-2-carboxylate
Formula:
C₇H₉NO₂ Peso molecolare: 139,16 g/mol Punto di ebollizione: 235°C |
MDL Number:
MFCD00817049 CAS Number: 2199-43-1 |
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 39.0 to 43.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...