Stampa…

1,1,2,3,3,3-Hexafluoropropyl ethyl ether ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Ethyl 1,1,2,3,3,3-Hexafluoropropyl ether

E1020-5G E1020-25G
TCIAE1020-5GEA 179 EUR
TCIAE1020-5G TCIAE1020-25G
1,1,2,3,3,3-Hexafluoropropyl ethyl ether ≥98.0% (by GC)
1,1,2,3,3,3-Hexafluoropropyl ethyl ether
Formula: C₅H₆F₆O
Punto di ebollizione: 65°C
MDL Number: MFCD00236122
CAS Number: 380-34-7
UN: 3271
ADR: 3,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR