Ethyl-1,5-Dimethylpyrazole-3-carboxylate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Ethyl 1,5-dimethyl-1H-pyrazole-3-carboxylate, 1,5-Dimethylpyrazole-3-carboxylic Acid Ethyl Ester
E1002-200MG
E1002-1G
TCIAE1002-200MGEA
31
EUR
TCIAE1002-200MG
TCIAE1002-1G
Ethyl-1,5-Dimethylpyrazole-3-carboxylate ≥98.0% (by GC)
Ethyl-1,5-Dimethylpyrazole-3-carboxylate
Formula:
C₈H₁₂N₂O₂ Peso molecolare: 168,20 g/mol Punto di ebollizione: 119°C |
MDL Number:
MFCD00085071 CAS Number: 5744-51-4 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 43.0 to 47.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...