Stampa…

Ethyl 3-(3,4-Dichlorophenyl)carbazate ≥98.0% (by GC)

Fornitore: TCI

Synonyms: 1-(3,4-Dichlorophenyl)-2-(ethoxycarbonyl)hydrazine, 3-(3,4-Dichlorophenyl)carbazic acid Ethyl ester

E0998-1G E0998-5G
TCIAE0998-1GEA 55 EUR
TCIAE0998-1G TCIAE0998-5G
Ethyl 3-(3,4-Dichlorophenyl)carbazate ≥98.0% (by GC)
Ethyl 3-(3,4-Dichlorophenyl)carbazate
Formula: C₉H₁₀Cl₂N₂O₂
MDL Number: MFCD28975114
CAS Number: 13124-15-7

Visualizza i codici

Risultati del test di specificazione

Purity (GC) min. 98.0 %
Melting point 110.0 to 114.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR