Stampa…

Ethyl (Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ethyl (Z)-2-Amino-alpha-(methoxyimino)-4-thiazoleacetate, (Z)-2-Amino-alpha-(methoxyimino)-4-thiazoleacetic acid Ethyl ester, (Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetic acid Ethyl ester

E0990-5G E0990-25G
TCIAE0990-5GEA 39 EUR
TCIAE0990-5G TCIAE0990-25G
Ethyl (Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetate ≥98.0% (by HPLC, titration analysis)
Ethyl (Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetate
Formula: C₈H₁₁N₃O₃S
Peso molecolare: 229,25 g/mol
MDL Number: MFCD00071532
CAS Number: 64485-88-7
EINECS: 264-912-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 162.0 to 166.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR