Stampa…

Ethyl methoxyacetate ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: Methoxyacetic ethyl ester

E0979-5G E0979-25G
TCIAE0979-5GEA 41 EUR
TCIAE0979-5G TCIAE0979-25G
Ethyl methoxyacetate ≥98.0% (by GC)
Ethyl methoxyacetate
Formula: C₅H₁₀O₃
Punto di ebollizione: 158°C
Flash Pt: 46°C
MDL Number: MFCD00009176
CAS Number: 3938-96-3
EINECS: 223-521-7
UN: 3272
ADR: 3,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR