Ethyl-2-bromoisovalerate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-Bromoisovaleric Acid Ethyl Ester, 2-Bromo-3-methylbutyric Acid Ethyl Ester, Ethyl 2-bromo-3-methylbutanoate, Ethyl 2-Bromo-3-methylbutyrate
E0969-25G
E0969-5G
TCIAE0969-25GEA
90
EUR
TCIAE0969-25G
TCIAE0969-5G
Ethyl-2-bromoisovalerate ≥98.0% (by GC)
Ethyl-2-bromoisovalerate
Formula:
C₇H₁₃BrO₂ Peso molecolare: 209,08 g/mol Punto di ebollizione: 186°C Flash Pt: 65°C |
MDL Number:
MFCD00026855 CAS Number: 609-12-1 EINECS: 210-178-3 UN: 3265 ADR: 8,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...