1-Ethynyl-4-(phenylethynyl)benzene ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-(4-Ethynylphenyl)-2-phenylacetylene, 4-Ethynyldiphenylacetylene
E0967-200MG
E0967-1G
TCIAE0967-200MGEA
67
EUR
TCIAE0967-200MG
TCIAE0967-1G
1-Ethynyl-4-(phenylethynyl)benzene ≥98.0% (by GC)
1-Ethynyl-4-(phenylethynyl)benzene
Formula:
C₁₆H₁₀ Peso molecolare: 202,26 g/mol |
MDL Number:
MFCD27665710 CAS Number: 92866-00-7 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 88.0 to 92.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...