Stampa…

(S)-Citalopram oxalate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (S)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile Oxalate

E0958-200MG E0958-1G
TCIAE0958-200MGEA 49 EUR
TCIAE0958-200MG TCIAE0958-1G
(S)-Citalopram oxalate ≥98.0% (by HPLC, titration analysis)
(S)-Citalopram oxalate
Formula: C₂₀H₂₁FN₂O
Peso molecolare: 414,43 g/mol
Punto di fusione: 146°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD06407826
CAS Number: 219861-08-2
Merck Index: 14,02318

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +12.0 to +14.0 deg (C=1,MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR