Econazole nitrate ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-[2-(4-Chlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]imidazole Nitrate, Ecostatin, 1-[2-(4-Chlorophenylmethoxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole nitrate
E0957-5G
E0957-25G
TCIAE0957-5GEA
52
EUR
TCIAE0957-5G
TCIAE0957-25G
Econazole nitrate ≥98.0% (by HPLC, titration analysis)
Econazole nitrate
Formula:
C₁₈H₁₅Cl₃N₂O·HNO₃ Peso molecolare: 444,69 g/mol |
MDL Number:
MFCD00058160 CAS Number: 24169-02-6 EINECS: 246-053-5 Merck Index: 14,03502 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...