Ethyl indole-3-carboxylate ≥97.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₁H₁₁NO₂ |
MDL Number:
MFCD00228454 CAS Number: 776-41-0 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 121.0 to 125.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...