Imidazole-2-carboxylic acid ethyl ester ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₆H₈N₂O₂ Punto di ebollizione: 51°C |
MDL Number:
MFCD03426031 CAS Number: 33543-78-1 |
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 179.0 to 183.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...