Ebselen ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₃H₉NOSe Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00210937 CAS Number: 60940-34-3 UN: 3077 ADR: 9,III Merck Index: 14,03486 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 181.0 to 185.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...