Stampa…

Ethyl-2-bromo-4-methylthiazole-5-carboxylate ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ethyl 2-bromo-4-methylthiazole-5-carboxylate

E0945-1G E0945-5G
TCIAE0945-1GEA 35 EUR
TCIAE0945-1G TCIAE0945-5G
Ethyl-2-bromo-4-methylthiazole-5-carboxylate ≥97.0% (by GC)
Ethyl-2-bromo-4-methylthiazole-5-carboxylate
Formula: C₇H₈BrNO₂S
Peso molecolare: 250,12 g/mol
MDL Number: MFCD03791227
CAS Number: 22900-83-0

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 97.0 %
Melting point 66.0 to 70.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR