1-Ethynylpyrene ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₈H₁₀ |
MDL Number:
MFCD02093933 CAS Number: 34993-56-1 |
Risultati del test di specificazione
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 115.0 to 119.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...