Stampa…

Ebastine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 4-(4-(Benzhydryloxy)piperidin-1-yl)-1-(4-(tert-butyl)phenyl)butan-1-one

E0925-5G E0925-1G
TCIAE0925-5GEA 246 EUR
TCIAE0925-5G TCIAE0925-1G
Ebastine ≥98.0% (by HPLC, titration analysis)
Ebastine
Formula: C₃₂H₃₉NO₂
Peso molecolare: 469,67 g/mol
MDL Number: MFCD00865661
CAS Number: 90729-43-4
Merck Index: 14,03484

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 85.0 to 89.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR